EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Cp*Rh-based indicator-displacement assays for the identification of amino sugars and aminoglycosides.
PMID 16521137
Abstract
Indicator-displacement assays based on the organometallic complex [{Cp*RhCl2}2] (Cp*=pentamethylcyclopentadienyl) and the dye gallocyanine were used to sense amino sugars and aminoglycosides in buffered aqueous solution by conducting UV-visible spectroscopy. The data of three assays at pH 7.0, 8.0, and 9.0 were sufficient to distinguish between the amino sugars galactosamine, glucosamine, mannosamine and the aminoglycosides kanamycin A, kanamycin B, amikacin, apramycin, paromomycin, and streptomycin. Furthermore, the assays were used to characterize mixtures of aminoglycosides and obtain quantitative information about the respective analytes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
124508
Gallocyanine, Dye content 90 %
C15H13ClN2O5