EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Terminal group effects on the fluorescence spectra of europium(III) nitrate complexes with a family of amide-based 2,3-dihydroxynaphthalene derivatives.
PMID 16524760
Abstract
Three ligands 2,2'-[2,3-naphthylenebis(oxy)]-bis(N,N-diethyl(acetamide)) (L(a)), 2,2'-[2,3-naphthylenebis(oxy)]-bis(N,N-diisopropyl(acetamide)) (L(b)) and 2,2'-[2,3-naphthylenebis(oxy)]-bis(N,N-dibutyl(acetamide)) (L(c)) and their europium(III) nitrate complexes were synthesized. The complexes were characterized by elemental analysis, IR, fluorescence spectroscopy and conductivity. The europium atoms are coordinated by O-atoms from CO, Ar-O-C. With the difference of the ligands, the solid fluorescent intensities of the Eu complexes vary regularly. Some factors that influencing the fluorescent intensity were discussed.
Related Materials