EMAIL THIS PAGE TO A FRIEND
Molecular diversity
A one-pot entry to a novel 3-H-benzo[d]pyrazolo[1,5-b]isothiazole ring system.
PMID 16528601
Abstract
The phosphorus ylide obtained from the reaction between 2-aminobenzo[d]isothiazol-3-one and dimethyl acetylenedicarboxylate in the presence of triphenylphosphine undergoes a smooth intramolecular Wittig-type reaction to produce, in a one-pot reaction, 3-H-benzo[d]pyrazolo[1,5-b]isothiazole-2,3a-dicarboxylic acid dimethyl ester, a novel functionalized heterocyclic compound.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D138401
Dimethyl acetylenedicarboxylate, 99%
C6H6O4