EMAIL THIS PAGE TO A FRIEND
FDA consumer
New warning on birth control patch.
PMID 16532540
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E1001
Ethisterone, ≥99% (HPLC)
C21H28O2
46272
Ethisterone, VETRANAL, analytical standard
C21H28O2