EMAIL THIS PAGE TO A FRIEND
Applied and environmental microbiology
Purification and characterization of two extracellular alkaline phosphatases from a psychrophilic arthrobacter isolate.
PMID 16535659
Abstract
Two extracellular, heat-labile alkaline phosphatases were purified from a psychrophilic Arthrobacter isolate, D10. The enzymes were active over different pH ranges, used distinct substrates, and had different kinetic properties. Each enzyme reacted specifically to its own antibody during immunoblot analysis. One had both monophosphatase and diesterase activities.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R0503 Reactive Red 120−Agarose, saline suspension, Type 3000-CL