EMAIL THIS PAGE TO A FRIEND
The Journal of chemical physics
Simple estimation of absolute free energies for biomolecules.
PMID 16542066
Abstract
One reason that free energy difference calculations are notoriously difficult in molecular systems is due to insufficient conformational overlap, or similarity, between the two states or systems of interest. The degree of overlap is irrelevant, however, if the absolute free energy of each state can be computed. We present a method for calculating the absolute free energy that employs a simple construction of an exactly computable reference system which possesses high overlap with the state of interest. The approach requires only a physical ensemble of conformations generated via simulation and an auxiliary calculation of approximately equal central-processing-unit cost. Moreover, the calculations can converge to the correct free energy value even when the physical ensemble is incomplete or improperly distributed. As a "proof of principle," we use the approach to correctly predict free energies for test systems where the absolute values can be calculated exactly and also to predict the conformational equilibrium for leucine dipeptide in implicit solvent.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L2752
Leu-Leu
C12H24N2O3