EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Fluoxetine up-regulates expression of cellular FLICE-inhibitory protein and inhibits LPS-induced apoptosis in hippocampus-derived neural stem cell.
PMID 16545775