EMAIL THIS PAGE TO A FRIEND
Appetite
Sensitivity to allyl isothiocyanate, dimethyl trisulfide, sinigrin, and cooked cauliflower consumption.
PMID 16546295
Abstract
The consumption of cauliflower consumers has been related to the olfactory and gustatory sensitivities to potentially objectionable flavor compounds in this vegetable. Based on the ascending concentration series method of limits, a first experiment was designed to develop rapid tests dedicated to estimate individuals' olfactory thresholds for allyl isothiocyanate (AITC) and dimethyl trisulfide (DMTS) and gustatory thresholds for sinigrin (SIN). The best compromise between rapidity and reliability was obtained with two replications of a four-alternative forced choice (AFC) at six ascending concentrations (6x2x4-AFC) for both AITC and DMTS, and with a 6x1x4-AFC for SIN. In a second experiment, sensitivity to SIN, AITC and DMTS was determined on 267 participants divided into three cauliflower consumer target groups: non-, medium- or high consumers. The non-consumers were significantly more sensitive to SIN and AITC than were the medium and high consumers. No effect of consumer's sensitivity to DMTS was observed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W327506
Dimethyl trisulfide, ≥98%, FG
C2H6S3
79592
Dimethyl trisulfide, analytical standard
C2H6S3