EMAIL THIS PAGE TO A FRIEND
Vestnik khirurgii imeni I. I. Grekova
[Extravesical treatment of the ureter and endoscopic correction of reflux in children].
PMID 1654647
Abstract
The authors have suggested the presence of reflux regulating the relationship between the urinary bladder and vesicoureteral segment which was called the vesico-osteal reflex (VOR). An original method of diagnostics of VOR with the help of profilometry was developed. A method of endoscopic correction of reflux was proposed with the help of glue MK-2 and KL-3 in combination with metoclopramide++ which made the treatment considerably more effective.