EMAIL THIS PAGE TO A FRIEND
Dalton transactions (Cambridge, England : 2003)
Aqueous rhodium(III) hydrides and mononuclear rhodium(II) complexes.
PMID 16547532
Abstract
In aqueous solutions, as in organic solvents, rhodium hydrides display the chemistry of one of the three limiting forms, i.e. {Rh(I)+ H+}, {Rh(II)+ H.}, and {Rh(III)+ H-}. A number of intermediates and oxidation states have been generated and explored in kinetic and mechanistic studies. Monomeric macrocyclic rhodium(II) complexes, such as L(H2O)Rh2+ (L = L1 = [14]aneN4, or L2 = meso-Me6[14]aneN4) can be generated from the hydride precursors by photochemical means or in reactions with hydrogen atom abstracting agents. These rhodium(II) complexes are oxidized rapidly with alkyl hydroperoxides to give alkylrhodium(III) complexes. Reactions of Rh(II) with organic and inorganic radicals and with molecular oxygen are fast and produce long-lived intermediates, such as alkyl, superoxo and hydroperoxo complexes, all of which display rich and complex chemistry of their own. In alkaline solutions of rhodium hydrides, the existence of Rh(I) complexes is implied by rapid hydrogen exchange between the hydride and solvent water. The acidity of the hydrides is too low, however, to allow the build-up of observable quantities of Rh(I). Deuterium kinetic isotope effects for hydride transfer to a macrocyclic Cr(v) complex are comparable to those for hydrogen atom transfer to various substrates.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF37380089
Rhodium, wire reel, 0.2m, diameter 0.25mm, hard, 99.9%
Rh
GF44569907
Rhodium, foil, 25x25mm, thickness 0.125mm, as rolled, 99.9%
Rh
GF80841146
Rhodium, foil, 25x25mm, thickness 0.15mm, as rolled, 99.9%
Rh
GF40539405
Rhodium, foil, 25x25mm, thickness 0.1mm, as rolled, 99.9%
Rh
GF20569697
Rhodium, foil, 12mm disks, thickness 0.5mm, as rolled, 99.9%
Rh
GF36379345
Rhodium, foil, light tested, 15x15mm, thickness 0.05mm, as rolled, 99.9%
Rh
GF57675561
Rhodium, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.9%
Rh
GF12568013
Rhodium, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, x-ray quality, 99.9%
Rh
GF96356559
Rhodium, foil, light tested, 25x25mm, thickness 0.075mm, as rolled, 99.9%
Rh
GF04978963
Rhodium, foil, not light tested, 10x10mm, thickness 0.012mm, as rolled, 99.9%
Rh
GF17514312
Rhodium, foil, not light tested, 25x25mm, thickness 0.00025mm, permanent polyester support, 99.9%
Rh
GF35442727
Rhodium, foil, not light tested, 25x25mm, thickness 0.0005mm, permanent polyester support, 99.85%
Rh
GF49036204
Rhodium, foil, not light tested, 25x25mm, thickness 0.00075mm, permanent polyester support, 99.85%
Rh
GF87742659
Rhodium, foil, not light tested, 25x25mm, thickness 0.001mm, permanent polyester support, 99.85%
Rh
GF15499413
Rhodium, foil, not light tested, 25x25mm, thickness 0.006mm, as rolled, 99.9%
Rh
GF24911726
Rhodium, foil, not light tested, 25x25mm, thickness 0.012mm, as rolled, 99.9%
Rh
GF82366342
Rhodium, foil, not light tested, 50x50mm, thickness 0.00025mm, permanent polyester support, 99.9%
Rh
GF22634120
Rhodium, foil, not light tested, 50x50mm, thickness 0.00075mm, permanent polyester support, 99.85%
Rh
GF33789550
Rhodium, foil, not light tested, 50x50mm, thickness 0.001mm, permanent polyester support, 99.85%
Rh
GF81993910
Rhodium, foil, not light tested, 50x50mm, thickness 0.006mm, as rolled, 99.9%
Rh
GF71687357
Rhodium, microfoil, disks, 10mm, thinness 0.1μm, specific density 124μg/cm2, permanent mylar 3.5μm support, 99.9%
Rh
GF94930667
Rhodium, microfoil, disks, 10mm, thinness 0.25μm, specific density 310μg/cm2, permanent mylar 3.5μm support, 99.99%
Rh
GF49946417
Rhodium, microfoil, disks, 10mm, thinness 0.50μm, specific density 620μg/cm2, permanent mylar 3.5μm support, 99.99%
Rh
GF14938005
Rhodium, microfoil, disks, 25mm, thinness 0.1μm, specific density 124μg/cm2, permanent mylar 3.5μm support, 99.9%
Rh
GF91583264
Rhodium, microfoil, disks, 25mm, thinness 0.25μm, specific density 310μg/cm2, permanent mylar 3.5μm support, 99.99%
Rh
GF64213680
Rhodium, wire reel, 0.05m, diameter 0.12mm, as drawn, 99.9%
Rh
GF45819995
Rhodium, wire reel, 0.05m, diameter 0.25mm, hard, 99.9%
Rh
GF74452316
Rhodium, wire reel, 0.05m, diameter 0.5mm, as drawn, 99.9%
Rh
GF83497652
Rhodium, wire reel, 0.05m, diameter 1.0mm, as drawn, 99.9%
Rh
GF26710680
Rhodium, wire reel, 0.1m, diameter 0.12mm, as drawn, 99.9%
Rh