EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Highly active, productive, and syndiospecific yttrium initiators for the polymerization of racemic beta-butyrolactone.
PMID 16548028
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
219126
β-Butyrolactone, 98%
C4H6O2