EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the USA
LHP1, the Arabidopsis homologue of HETEROCHROMATIN PROTEIN1, is required for epigenetic silencing of FLC.
PMID 16549797