EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Formation of superoxide by benzofurazans in Escherichia coli under aerobic incubation.
PMID 1655293
Abstract
The superoxide (O2-.) production in Escherichia coli through the action of benzofurazans (BZs) was examined using the cytochrome c (cyt. c) reduction method. Adding BZs to E. coli cell suspensions caused the cyt. c reduction, which was completely inhibited by superoxide dismutase (SOD). The effects of BZs on cyt. c reduction was in the order of benzofurazan (1) approximately 4,7-dimethylbenzofurazan (2) approximately 4,7-dibromobenzofurazan (3) less than 4-bromo-6-cyanobenzofurazan (4) less than 4,7-dicyanobenzofurazan (5). This was correlated with the toxicity of BZs against E. coli growth (1 approximately 2 approximately 3 less than 4 less than 5) and with the redox potentials of BZs (1 approximately 2 less than 3 less than 4 less than 5). The formation of compound 5 anion radical in the cell suspensions in the absence of dioxygen (O2), was determined using ESR spectrum. The ESR signal of the anion radical disappeared with the addition of O2. The BZs effected the O2-. production in E. coli cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
650137
Benzofurazan, 97%
C6H4N2O