EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
An efficient stereoselective dihydroxylation of glycals using a bimetallic system, RuCl3/CeCl3/NaIO4.
PMID 16555855
Abstract
A catalytic dihydroxylation reaction on glycals has been developed using a bimetallic oxidizing system to furnish sugar 1,2-diols in a highly stereoselective manner.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
208523
Ruthenium(III) chloride, Ru content 45-55%
Cl3Ru