EMAIL THIS PAGE TO A FRIEND
Organic letters
A versatile approach for the asymmetric syntheses of (1R,9aR)-epiquinamide and (1R,9aR)-homopumiliotoxin 223G.
PMID 16562910
Abstract
[reaction: see text] Using 5b as a common intermediate, the first asymmetric synthesis of (-)-epiquinamide (4) and a formal asymmetric synthesis of (-)-homopumiliotoxin 223G (2) is described. A key feature of our approach is the flexible introduction of a functionalized C(4) side chain to (S)-3-benzyloxyglutarimide 7 in a regio- and diastereoselective manner. Utilization of a tandem Swern oxidation-Grignard addition strategy efficiently prevented racemization. An unexpected NaN(3)-promoted methanesulfonic acid elimination yielded 17, a reaction which could be useful for the syntheses of 8-dehydrodesmethylpumiliotoxins such as alkaloid 235C (3).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
178098
Glutarimide, 98%
C5H7NO2