EMAIL THIS PAGE TO A FRIEND
Journal of environmental pathology, toxicology and oncology : official organ of the International Society for Environmental Toxicology and Cancer
An overview of synthetic approaches to porphyrin, phthalocyanine, and phenothiazine photosensitizers for photodynamic therapy.
PMID 16566711
Abstract
The mechanistic basis of photodynamic therapy is reviewed briefly, and the factors that are important in improving photosensitizers are examined. The methods for preparing the three main classes of PDT photosensitizers, namely, porphyrins, phthalocyanines, and phenothiazines, are reviewed.