EMAIL THIS PAGE TO A FRIEND
Biodegradation
Biodegradation of hexazinone by two isolated bacterial strains (WFX-1 and WFX-2).
PMID 16570230
Abstract
Two hexazinone-degrading bacterial strains were isolated from soil by enrichment culture technique, and identified as Pseudomonas sp. and Enterobacter cloacap, respectively. The two purified isolates, designated as WFX-1 and WFX-2, could rapidly degrade hexazinone with half-lives of 3.08 and 2.95 days in mineral salts medium (hereafter referred to as MSM). In contrast, their mixed bacterial culture (herein abbreviated as MBC) was found to degrade hexazinone, at an initial concentration of 50 mg l(-1), by enhancing 2.3-fold over that when the isolates were used alone. The degradation of hexazinone by MBC in MSM clearly decreased concomitant with the increase of initial concentration, and the level of hexazinone that was toxic enough to totally inhibit degradation was in the range of 150-200 mg l(-1). The appropriately combined conditions for hexazinone degradation by MBC in MSM were studied, and found to be pH 5.5, 30 degrees C and at agitation of 120 rpm. The addition of MBC to soil had a greater impact on disappearance of hexazinone, which nearly increased fivefold over that of the control set. As a result, findings in the present investigation provide useful information for soil and water decontamination of hexazinone.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36129
Hexazinone, PESTANAL®, analytical standard
C12H20N4O2