EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Studies on chemical constituents of the brown alga Dictyopteris divaricata].
PMID 16570798
Abstract
To investigate the chemical constituents of the brown alga D. divaricata, and to test cytotoxicities of the purified compounds. Compounds were isolated by normal phase silica gel, Sephadex LH-20 chromatography and reverse phase HPLC techniques. Their structures were elucidated by spectroscopic methods including IR, MS and NMR. Cytotoxicities were tested by MTT method. Eight compounds were isolated from ethanolic extract of the brown alga D. divaricata and their structures were identified as (-)-torreyol (I), 4beta, 5alpha-dihydroxycubenol (II), 3-farnesyl-p-hydroxybenzioc acid (III), chromazonarol (IV), fucosterol (V), phenyl acetylamine (VI), 4-hydroxybenzoic acid (VII) and n-hexadecanoic acid (VIII). Compound II and IV were obtained from this alga for the first time. The others were isolated from the Dictyotaceae algae for the first time. All compounds were inactive (IC50 > 10 microg x mL(-1)) against human tumor cell lines KB, Bel-7402, PC-3M, Ketr 3 and MCF-7.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F5379
Fucosterol, ≥93%
C29H48O