EMAIL THIS PAGE TO A FRIEND
Doklady biological sciences : proceedings of the Academy of Sciences of the USSR, Biological sciences sections
Humic substances as natural photoinducers degrading pesticides in the environment.
PMID 16572825