EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
Spectrophotometric determination of carbinoxamine maleate in pharmaceutical formulations by ternary complex formation with Cu(II) and eosin.
PMID 16574052
Abstract
A simple and sensitive spectrophotometric method was developed for the determination of carbinoxamine maleate in pharmaceutical formulations. The method is based on the formation of a ternary complex, extractable with chloroform, between copper(II), eosin, and carbinoxamine maleate. The absorption spectra of the ternary complexes shows, under optimum conditions, a maxima at 538 nm, with apparent molar absorptive 6.1690 x 10(4) mol(-1) cm(-1), Sandell's sensitives 6.75 x 10(-3) microg cm(-2), and linearity in the concentration range 0.75-10.0 microg ml(-1). The method can be achieved with high accuracy (recovery values, 100 +/- 2%) and precision (with standard deviation 0.029-0.155 and relative standard deviation 3.87-1.55%). The method was again successfully applied, with high accuracy and good precision, for the determination of carbinoxamine maleate in various pharmaceutical formulations (syrup, drops, and tablets).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8278
Carbinoxamine maleate salt, analytical standard
C16H19ClN2O · C4H4O4