EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Oligomers of cis-beta-norbornene amino acid: formation of beta-strand mimetics.
PMID 16575456
Abstract
The oligomers of constrained cis-exo-beta-norbornene amino acid were synthesised and characterised by extensive NMR, CD, IR and MD studies. The results showed the formation of both right and left handed consecutive 6-membered hydrogen-bonded strands for [2S,3R] and [2R,3S] enantiomers, respectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N32407
Bicyclo[2.2.1]hept-2-ene, 99%
C7H10