EMAIL THIS PAGE TO A FRIEND
FEBS letters
Improvement of hypertension and vascular dysfunction by hydroxyhydroquinone-free coffee in a genetic model of hypertension.
PMID 16579992
Abstract
Chlorogenic acid, a polyphenol found in coffee, has antihypertensive actions, but epidemiologic data on the effects of coffee on blood pressure are controversial. Specific coffee components that inhibit the hypotensive effect of chlorogenic acid and the physiologic mechanisms underlying the effects of coffee without these components were investigated. One component, hydroxyhydroquinone (HHQ), inhibited the hypotensive effects of chlorogenic acid in spontaneously hypertensive rats (SHR). The attenuation of hypertension by HHQ-free coffee was associated with nitric oxide, the suppression of mRNA expression of NAD(P)H oxidase, and the improvement in endothelium-dependent vasodilation in the aorta. Thus, HHQ-free coffee might regulate vascular tone by improving the bioavailability of nitric oxide in SHR.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
173401
1,2,4-Benzenetriol, ReagentPlus®, 99%
C6H6O3