EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Controlled one-step fabrication of highly oriented ZnO nanoneedle/nanorods arrays at near room temperature.
PMID 16583010
Abstract
Highly oriented ZnO nanoneedle/nanorods arrays have been fabricated by direct oxidation of zinc foil in alkaline zincate ion solution at near room temperature (20 degrees C for nanoneedles, 30 degrees C for nanorods).