EMAIL THIS PAGE TO A FRIEND
The Journal of pharmacology and experimental therapeutics
Pharmacologic actions of Ro 24-5913, a novel antagonist of leukotriene D4.
PMID 1658310
Abstract
Ro 24-5913, (E)-4-[3-[2-(4-cyclobutyl-2- thiazolyl)ethenyl]phenylamino]-2,2-diethyl-4-oxobutanoic acid, has been identified as a chemically unique, potent and selective LTD4 antagonist. In vitro, Ro 24-5913 competes with [3H]LTD4 for its binding site on guinea pig lung membranes with an IC50 of 6.4 +/- 2.2 nM. In isolated guinea pig tracheal smooth muscle, Ro 24-5913 produces concentration-dependent rightward shifts of LTD4-induced contraction curves (pA2 value of 9.6 +/- 0.2). The slope of the Schild plot is not significantly different from 1, indicating that the antagonism is of a competitive nature. In the human bronchus, Ro 24-5913 is an effective antagonist of LTD4-induced contractions (pKB of 9.3 +/- 0.1). In vivo, Ro 24-5913 dose-dependently inhibits LTD4-induced bronchoconstriction in guinea pigs by the i.v. (ID50 0.13 mg/kg), oral (ID50 0.12 mg/kg) and aerosol (IC50 0.008%) routes of administration. This in vivo activity is specific as evidenced by the inability of Ro 24-5913 to inhibit bronchoconstriction induced by LTB4, PAF or histamine. In comparison with other LTD4 antagonists evaluated in this guinea pig model, Ro 24-5913 is markedly superior in terms of oral potency, bioavailability and oral duration of action. Ro 24-5913 also blocks allergic bronchospasm mediated by endogenously generated leukotrienes in guinea pigs; the potency and duration of action is nearly equivalent to that seen as an antagonist of bronchoconstrictions produced by exogenous LTD4. In summary, Ro 24-5913 is representative of a novel chemical class of LTD4 receptor antagonists.(ABSTRACT TRUNCATED AT 250 WORDS)
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C6239
Cinalukast, ~98% (HPLC)
C23H28N2O3S