EMAIL THIS PAGE TO A FRIEND
The Lancet. Oncology
Xaliproden lessens oxaliplatin-mediated neuropathy.
PMID 16598880
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
X4879
Xaliproden hydrochloride, ≥97% (HPLC), solid
C24H22F3N · HCl