EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Synthesis and structure revision of calyxin natural products.
PMID 16599616
Abstract
Tandem Prins cyclization and Friedel-Crafts reaction with an electron-rich aromatic ring were used to prepare the core structures of calyxin natural products. The proposed structure of epicalyxin F was prepared and shown to be incorrect. Several calyxin natural products, including calyxin F and L, were synthesized, and the structures were reassigned on the basis of NMR data and synthetic correlations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
36199
Tridemorph, PESTANAL®, analytical standard
C19H39NO