EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Regioselective copper-catalyzed amination of bromobenzoic acids using aliphatic and aromatic amines.
PMID 16599627
Abstract
A chemo- and regioselective copper-catalyzed cross-coupling procedure for amination of 2-bromobenzoic acids is described. The method eliminates the need for acid protection and produces N-aryl and N-alkyl anthranilic acid derivatives in up to 99% yield. N-(1-Pyrene)anthranilic acid has been employed in metal ion-selective fluorosensing. Titration experiments showed that this pyrene-derived amino acid forms an equimolar complex with Hg(II) in water resulting in selective fluorescence quenching even in the presence of other metal ions such as Zn(II) and Cd(II).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
560219
2-Bromo-3-methylbenzoic acid, 97%
C8H7BrO2
367931
2-Bromo-4-fluorobenzoic acid, 97%
C7H4BrFO2