EMAIL THIS PAGE TO A FRIEND
Zhurnal vysshei nervnoi deiatelnosti imeni I P Pavlova
[A stimulated shift in heart rhythm stability as a condition for CNS activation and stimulus perception].
PMID 1660664
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D98807
N,N-Diethylnicotinamide, 99%
C10H14N2O