EMAIL THIS PAGE TO A FRIEND
Archives of neurology
Effects of anticonvulsant drugs on life span.
PMID 16606760
Abstract
Aging is characterized by widespread degenerative changes in tissue morphology and function and an increase in the incidence of human diseases such as cancer, stroke, and Alzheimer disease. Findings from recent genetic studies suggest that molecular mechanisms that influence life span are evolutionarily conserved, and interventions that extend the life span of model organisms such as worms and flies are likely to have similar effects on vertebrates such as humans. However, little progress has been made in identifying drugs that delay aging. We identified 3 pharmacologic compounds, ethosuximide, trimethadione, and 3,3-diethyl-2-pyrrolidinone, that extend lifespan and delay age-related degenerative changes in the nematode worm Caenorhabditis elegans. All 3 compounds are anticonvulsants that modulate neural activity in vertebrates, and ethosuximide and trimethadione are used to treat absence seizures in humans. We discuss existing evidence that these drugs might also delay vertebrate aging and suggest experiments that could test this hypothesis. Genetic and cell ablation studies conducted with model organisms have demonstrated connections between the nervous system and aging. Our studies provide additional support for the hypothesis that neural activity plays a role in lifespan determination, since ethosuximide and trimethadione regulated neuromuscular activity in nematodes. Our findings suggest that the lifespan extending activity of these compounds is related to the anticonvulsant activity, implicating neural activity in the regulation of aging. We also discuss models that explain how the nervous system influences lifespan.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T0781
3,5,5,-Trimethyloxazolidine-2,4-dione, analytical standard
C6H9NO3