EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
A quinhydrone-type 2 ratio 1 acceptor-donor charge transfer complex obtained via a solvent-free reaction.
PMID 16609793
Abstract
A 2 : 1 2-methoxybenzoquinone (BQ(OMe))-hydroquinone (H2Q) complex (5), obtained as single crystals by a solvent-free reaction, combines dipolar and pi-pi interactions, as well as hydrogen bonding.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
282960
Quinhydrone, 97%
C6H6O2 · C6H4O2