EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
A lag-phase in the reduction of flavin dependent thymidylate synthase (FDTS) revealed a mechanistic missing link.
PMID 16609803
Abstract
An unexpected substrate-dependent lag-phase, found in the single turnover reduction of FDTS bound flavin, sheds light on the molecular mechanism of this alternative thymidylate synthase.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D3876
2′-Deoxyuridine 5′-monophosphate disodium salt, Sigma Grade
C9H11N2O8P · 2Na