EMAIL THIS PAGE TO A FRIEND
Skin pharmacology and physiology
Establishing a minimal erythema concentration of methyl nicotinate for optimum evaluation of anti-inflammatories.
PMID 16612142
Abstract
Topical administration of chemicals such as methyl nicotinate that induce erythema have been employed to measure the effectiveness of formulations containing anti-inflammatory agents. Prior studies have utilized a single concentration of methyl nicotinate, between 36.5 and 100 mM, for all test subjects in evaluations of topical formulations. However, individuals have different thresholds of response to methyl nicotinate; thus, a single concentration may not be appropriate for all individuals and could result in the apparent lack of anti-inflammatory activity of the formulation being evaluated. In the current study, we evaluated the use of a minimal erythema concentration (MEC) of methyl nicotinate, defined as the lowest concentration that produces a complete and even erythema at the test site, compared with a 36.5-mM concentration of methyl nicotinate. Hydroalcoholic gels containing the nonsteroidal anti-inflammatory drug ibuprofen were compared with placebo. Diffuse reflectance spectroscopy was employed to measure differences in cutaneous inflammatory response between the control (placebo)-treated group and the ibuprofen-treated group. When chemical erythema was induced using an MEC of methyl nicotinate, greater reductions in erythema were seen in ibuprofen-treated sites compared with sites treated with a 36.5-mM concentration of methyl nicotinate. In conclusion, for an accurate assessment method of erythema induced by methyl nicotinate, consideration should be given to determining the extent of response of an erythema-producing agent on an individual basis. An MEC of methyl nicotinate should be determined and employed for each individual to obtain more consistent and reliable efficacy results of anti-inflammatory activity.