EMAIL THIS PAGE TO A FRIEND
Endocrinology
Insulin-like growth factor binding protein 3 has opposing actions on malignant and nonmalignant breast epithelial cells that are each reversible and dependent upon cholesterol-stabilized integrin receptor complexes.
PMID 16614079
Abstract
IGF-binding protein (IGFBP)-3 is generally considered to have actions that counterbalance those of IGFs and is therefore being developed as a cancer treatment. In breast tumors, however, high levels are associated with aggressive tumors and poor prognosis. Consistent with this we have demonstrated that although IGFBP-3 and a non-IGF-binding fragment (serine phosphorylation domain peptide) reduced attachment and enhanced apoptosis of Hs578T breast cancer cells cultured on collagen or laminin, it promoted their attachment and survival on fibronectin, which is abundant in the matrix of aggressive tumors. We have now examined the factors that determine whether IGFBP-3 has positive or negative actions on breast epithelial cells. IGFBP-3 also promoted survival of Hs578T cells in the presence of an antibody to the beta1-integrin subunit or when cholesterol-stabilized complexes were disrupted. These actions were blocked by IGF-I or a MAPK inhibitor. Serine phosphorylation domain peptide had similar actions on MCF-7 cells that were again reversed on fibronectin or with disruption of cholesterol-stabilized complexes and blocked by the beta1-integrin antibody. In contrast, IGFBP-3 promoted growth and survival for nonmalignant MCF-10A cells, but these effects were again reversed on fibronectin and blocked by the beta1 antibody or a MAPK inhibitor or by disruption of cholesterol-stabilized complexes. On Hs578T cells, IGFBP-3 bound to caveolin-1 and beta1-integrins, enhancing their aggregation, the recruitment of focal adhesion kinase, and the activation of MAPK. In summary, with three breast epithelial cell lines, IGFBP-3 had positive or negative effects on growth and survival dependent upon the status of cholesterol-stabilized integrin receptor complexes.