EMAIL THIS PAGE TO A FRIEND
The Journal of investigative dermatology
Functioning methionine-S-sulfoxide reductases A and B are present in human skin.
PMID 16619011
Abstract
Methionine residues in the structure of proteins and peptides are especially sensitive to oxidation by hydrogen peroxide (H(2)O(2)) yielding both the (R) and (S) diastereomers of methionine sulfoxide. This commentary shows that both diastereomers of methionine sulfoxide (R and S) can be repaired in the human epidermis by methionine sulfoxide reductases A and B, respectively.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
64430
DL-Methionine sulfoxide, ≥99.0% (NT)
C5H11NO3S