EMAIL THIS PAGE TO A FRIEND
Journal of chromatographic science
Determination and characterization of the pyrolysis products of isoprocarb by GC-MS.
PMID 16620509
Abstract
The pyrolysis behavior of isoprocarb (an insecticide with contact and stomach action) is investigated using pyrolysis-gas chromatography-mass spectrometry. The pyrolysis products are separated using an HP-5 column under temperature program with helium as the carrier gas. The total of 80 separated pyrolysis components at 600 degrees C, 750 degrees C, and 900 degrees C under helium atmosphere are identified using a probability-based matching search procedure, combined with the correlation of boiling point (BP) and Lee retention index (RI). Some of the BP values of the tentative components are estimated using the group contributions method because experimental values are not available. The levels of the identified components are estimated by the peak area normalization method from the chromatogram. It is found that isoprocarb decomposes more with the increase of temperature, and a large number of mono aromatics and polycyclic aromatic hydrocarbons and their derivatives are produced when the pyrolysis temperature is higher than 750 degrees C. The content of the decomposition products in the pyrolysate varies from 0.04% to 22.20%.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45541
Isoprocarb, PESTANAL®, analytical standard
C11H15NO2