EMAIL THIS PAGE TO A FRIEND
Colloids and surfaces. B, Biointerfaces
Cationic liposomes modified with non-ionic surfactants as effective non-viral carrier for gene transfer.
PMID 16626948
Abstract
A defined change in formulation components affects the physical and chemical characteristics of cationic liposomes (CLs) carriers in many ways. Therefore, a great degree of control can be exercised over the structure by modifying the CLs with various materials, leading to new innovations for carrier improvement. In the present study, surface modifications of cationic liposomes with non-ionic surfactants--sorbitan monoesters serials (Span 85, 80, 40 and 20) were carried out for developing a new gene transfer carrier. Span modified cationic liposomes (Sp-CLs) were prepared by reverse phase evaporation method (RPV) and self-assemble complexes of antisense oligonucleotides/surfactant modifying cationic liposomes were prepared by auto-coacervation through electrostatic effect. Characterization of Sp-CLs and the self-assembled complex was performed by electron microscope, particle size, zeta potential, turbidity and agarose electrophoresis. Furthermore, in vitro cellular uptake experiment showed that Span plays a role in enhancing the cellular uptake of encapsulated oligonucleotides mediated by Sp-CLs by the endocytosis-dependent route. CLs modified with Span 40 significantly facilitated the cellular uptake by COS-7 cells and HeLa cells; also showed some positive effect on gene expression. That suggests it is a potential non-viral carrier for efficient gene transfer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
388920
Sorbitan monopalmitate
C22H42O6
S6635
Span® 20
S7135
Span® 85