EMAIL THIS PAGE TO A FRIEND
Communications in agricultural and applied biological sciences
Multi-residue method for determination of pesticides residues in potatoes by GC-NPD.
PMID 16628959
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45393
Chlorpropham, PESTANAL®, analytical standard
C10H12ClNO2