EMAIL THIS PAGE TO A FRIEND
European journal of pharmacology
Effects of adenosine A(3) receptor agonist on bone marrow granulocytic system in 5-fluorouracil-treated mice.
PMID 16643889
Abstract
The purpose of the experiments reported was to investigate effects of N(6)-(3-iodobenzyl)adenosine-5'-N-methyluronamide (IB-MECA), a selective adenosine A(3) receptor agonist, on the granulocytic system in femoral marrow of mice depleted by the cytotoxic drug 5-fluorouracil. In the phase of the highest cell depletion IB-MECA was injected i.p. at single doses of 200 nmol/kg given either once or twice daily in 2- and 4-day regimens starting on day 1 after 5-fluorouracil administration; the effects were evaluated on days 3 and 5, respectively. The general effect of IB-MECA in all these experiments was an enhancement of the counts of morphologically recognizable proliferative granulocytic cells, interpreted as evidence of the differentiation of committed progenitor cells. A more expressive effect was observed after IB-MECA injected twice daily. It was found that the induction of the strong differentiation pressures by IB-MECA given twice daily shortly after 5-fluorouracil treatment can be counterproductive due to the preponderance of differentiaton processes over the proliferation control. In additional experiments, it has been shown that the use of the 2-day administration of IB-MECA given twice daily in the recovery phase, i.e., on days 5 and 6 after 5-fluorouracil administration, does not induce stimulatory effects. Thus, the dosing and timing of IB-MECA treatment determines its effectivity in stimulating granulopoiesis under conditions of myelosuppression.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I146
IB-MECA, solid, ≥98% (HPLC)
C18H19IN6O4