EMAIL THIS PAGE TO A FRIEND
European journal of pediatrics
Hypersensitivity to racecadotril: a case report.
PMID 16649028
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0043
Racecadotril, ≥98% (HPLC)
C21H23NO4S