EMAIL THIS PAGE TO A FRIEND
Proceedings of the Western Pharmacology Society
Comparison of the cardiotonic response of ouabain, androstan-3-one, 16-dehydroprogesterone, and conjugated estrogens.
PMID 1664956