EMAIL THIS PAGE TO A FRIEND
Plant physiology
Electrophoretic transfer as a technique for the detection and identification of plant amylolytic enzymes in polyacrylamide gels.
PMID 16663595
Abstract
An electrophoretic transfer technique was developed for the specific identification of isozymes of starch debranching enzyme, alpha-amylase, and beta-amylase. Amylolytic enzymes are separated by native polyacrylamide slab gel electrophoresis and proteins in gels are electrophoretically transferred through starch-containing polyacrylamide gels. Each amylolytic enzyme degrades starch in the transfer gel to its characteristic limit dextrin as it moves through the gel. Various limit dextrins in the starch gel are identified by their characteristic color development in KI/I solution. Isozymes of starch debranching enzyme, alpha-amylase, and beta-amylase can be easily identified in the same gel.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
93352
Trizma® base, ≥99.0% (T)
C4H11NO3
04577
Trizma® base, tested according to Ph.Eur.
C4H11NO3