EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
In(OTf)3-catalyzed tandem nucleophilic addition and cyclization of ortho-alkynylarylaldimines to 1,2-dihydroisoquinolines.
PMID 16671144
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
251062
2,6-Di-tert-butyl-4-methoxyphenol, 97%
C15H24O2