EMAIL THIS PAGE TO A FRIEND
Rapid communications in mass spectrometry : RCM
Investigation of non-covalent complexes of HIV-1 promoter DNA and polyamides containing N-methylpyrrole by electrospray ionization mass spectrometry.
PMID 16676310
Abstract
Eight novel polyamides containing N-methylpyrrole were designed to target the sequence (5'-CTGCATATAAGCAG-3'/5'-CTGCTTATATGCAG-3') of the TATA box element of the HIV-1 promoter DNA. The non-covalent complexes of the promoter DNA and the polyamides were investigated by electrospray ionization (ESI) mass spectrometry, which provided strong evidence for the binding of the novel polyamides to the sequence of the TATA box element. It also revealed that polyamide 2 (PyPyPyPybetaDp), a potent binder of HIV-1 promoter DNA and a lead molecule for the design of new anti-HIV-1 drugs, had the highest binding affinity with the TATA box element DNA among these polyamides by examining the stoichiometry and the selectivity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W509973
1-Methylpyrrole, ≥99%
C5H7N
M78801
N-Methylpyrrole, 99%
C5H7N