EMAIL THIS PAGE TO A FRIEND
Bioorganic & medicinal chemistry letters
Small-molecule microarrays: development of novel linkers and an efficient detection method for bound proteins.
PMID 16682190
Abstract
Novel isocyanate and diazoketone linkers possessing polyoxypropylenediamine as a spacer for small-molecule microrray are developed. White light interferometry is introduced to detect bound proteins on the glass slides without using chemically modified proteins.
Related Materials