EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Determination of ibuprofen and flurbiprofen in pharmaceuticals by capillary zone electrophoresis.
PMID 16687231
Abstract
Capillary zone electrophoresis with spectrophotometric detection was used for the determination of ibuprofen (IB) and flurbiprofen (FL) in pharmaceuticals. The separation was carried out in a fused silica capillary (60 cm x 100 microm i.d. effective length 45 cm) at 30 kV with UV detection at 232 nm. The optimized background electrolyte was 20mM N-(2-acetamido)-2-aminoethanesulfonic acid (ACES) with 20mM imidazole and 10mM alpha-cyclodextrin of pH 7.3. 2-Naphthoxyacetic acid was used as internal standard. A single analysis took less than 5 min. Rectilinear calibration ranges were 2-500 mg l(-1) for IB and 1-60 mg l(-1) for FL. The relative standard deviations (R.S.D.) values (n=6) were 1.53% for IB and 1.29% for FL (for 200 mg l(-1) IB and 10 mg l(-1) FL). This validated method has been successfully applied for the routine analysis of 10 commercially available pharmaceutical preparations (syrup, tablets, cream and gel).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
00194
ACES, BioUltra, ≥99.5% (T)
C4H10N2O4S