EMAIL THIS PAGE TO A FRIEND
Fiziologicheskii zhurnal SSSR imeni I. M. Sechenova
[The ratio of sex hormone production by the ovaries and the adrenal cortex in different phases of the rat estrous cycle].
PMID 1668743
Abstract
The biosynthesis of estradiol and testosterone was found to occur mainly in the ovaries (4-5 and 10-20 times more than in the adrenals during diestrus and proestrus, resp.). Estron and androstendion were produced equally in both glands. The estrogens and androgens were found to act as synergists in the stimulation of biosynthetic processes in the uterus. Their level in peripheral blood reflects less fully the character of hormonal function of the ovaries than study of these steroids in the incubates of the ovaries. The biosynthesis of progesterone in adrenals exceeds 2-3-fold that in the ovaries. No changes on production of this hormone depending on the cycle phase, were found.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A41909
4-Aminobenzoic hydrazide, 95%
C7H9N3O