EMAIL THIS PAGE TO A FRIEND
The Journal of General Virology
Envelope proteins of spleen necrosis virus form infectious human immunodeficiency virus type 1 pseudotype vector particles, but fail to incorporate upon substitution of the cytoplasmic domain with that of Gibbon ape leukemia virus.
PMID 16690921