EMAIL THIS PAGE TO A FRIEND
CDR (London, England : Weekly)
'Sheep flu'.
PMID 1669884
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
34371
Propetamphos, PESTANAL®, analytical standard
C10H20NO4PS