EMAIL THIS PAGE TO A FRIEND
Medical mycology
Tween 40-based precipitate production observed on modified chromogenic agar and development of biological identification kit for Malassezia species.
PMID 16702101
Abstract
We developed a simple identification kit for nine species of Malassezia (M. furfur, M. slooffiae, M. sympodialis, M. restricta, M. obtusa, M. globosa, M. pachydermatis, M. dermatis, and M. japonica) based on their biological features. This method utilizes Tween 40-based precipitate production on modified chromogenic agar (CHROMagar) Malassezia medium, growth on specific agars (Sabouraud's dextrose agar, Cremophor EL agar, Tween 60-esculin agar), and catalase reactions. This identification kit was verified with 11 type and reference strains of nine Malassezia species. An additional 26 clinical isolates were also successfully identified using the kit and the results were confirmed by molecular biological analysis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P1504 TWEEN® 40, viscous liquid