EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Functionalisation of carbon nanohorns.
PMID 16703130
Abstract
The functionalisation of single wall carbon nanohorns via 1,3-dipolar cycloaddition has been achieved, and the products have been characterised by spectroscopy, microscopy and thermogravimetry.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H2120
Heptaldehyde, 95%
C7H14O
W254002
Heptanal, ≥95%, FCC, FG
C7H14O